Learn More
Butabindide oxalate, Tocris Bioscience™
High affinity CCK-inactivating serine protease inhibitor
Brand:  Tocris (Bio-Techne) 132310
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13449143
  • £195.51 / 10mg
Estimated Shipment date
from Supplier 17-12-2019
Description
Description
High affinity, reversible, selective and competitive inhibitor of a CCK-inactivating serine protease (tripeptidyl peptidase II) (Ki = 7 nM). Active in vivo (ID50 = 1.1 and 6.8 mg/kg i.v. for inhibition of liver and brain enzyme respectively).
Specifications
Chemical Identifiers
C19H27N3O6
Specifications
Butabindide oxalate
>99%
393.44
185213-03-0
C19H27N3O6