Learn More
Butabindide oxalate, Tocris Bioscience™
High affinity CCK-inactivating serine protease inhibitor
POA
Specifications
CAS 185213-03-0
View More Specs
Description
Description
High affinity, reversible, selective and competitive inhibitor of a CCK-inactivating serine protease (tripeptidyl peptidase II) (Ki = 7 nM). Active in vivo (ID50 = 1.1 and 6.8 mg/kg i.v. for inhibition of liver and brain enzyme respectively).
Specifications
Specifications
185213-03-0