missing translation for 'onlineSavingsMsg'
Learn More

Millipore™ Bugbuster™ Master Mix

Brand:  Millipore™ 71456-4

Additional Details : Weight : 0.05000kg

Product Code. 15186166

  • £678.67 / 500mL


Estimated Shipment
27-06-2018
Add to basket

Specifications

Specifications

500mL
Bugbuster Master Mix
2 to 8°C
Videos