missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution pH9.2 (Borate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical™

£38.59 - £349.89

Chemical Identifiers

MDL Number 163147
View More Specs

Products 5
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10082531
SDS View Documents
Fisher Chemical
J/2870/08
500mL HDPE plastic bottle
£38.59
500mL
View alternative products
Save Up to 
»
In stock, ready to ship: 34
Please to purchase this item. Need a web account? Register with us today!
10142000
SDS View Documents
Fisher Chemical
J/2870/15
1L HDPE plastic bottle
£68.94
1 L
View alternative products
Save Up to 
»
In stock, ready to ship: 140
Please to purchase this item. Need a web account? Register with us today!
10590911
SDS View Documents
Fisher Chemical
J/2870/17
2.5L HDPE plastic bottle
£139.80
2.50 L
View alternative products
Save Up to 
»
In stock, ready to ship: 31
Please to purchase this item. Need a web account? Register with us today!
10030180
SDS View Documents
Fisher Chemical
J/2870/21
5L Plastic container
£191.63
5 L
View alternative products
Save Up to 
»
Estimated Shipment: 01-11-2021
Please to purchase this item. Need a web account? Register with us today!
10419370
SDS View Documents
Fisher Chemical
J/2870/24
10L Bag in box
£349.89
10 L
View alternative products
Save Up to 
»
In stock, ready to ship: 3
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Buffer Solution pH9.2 (Borate), NIST Standard Solution ready to use for pH measurement

Specifications

Chemical Identifiers

163147

Specifications

163147
HDPE plastic bottle
Colorless
Liquid
9.2
500mL
SDS
Product Certifications