missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution pH9.2 (Borate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical™

Brand:  Fisher Chemical J/2870/17

128.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.



Code : 23

Additional Details : CAS Number : 7732-18-5 Weight : 2.50000kg


 View more versions of this product

Product Code. 10590911

  • £203.00 / 2.50 L
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 

Description

Description

Buffer Solution pH9.2 (Borate), NIST Standard Solution ready to use for pH measurement

Specifications

Specifications

Colorless
Liquid
163147
9.2
2.5 L
HDPE plastic bottle
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Reproductive toxicity Category 1B

Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS Documents

Documents

Certificates
Special Offers

Special Offers