missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution pH9.2 (Borate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical™
Brand: Fisher Chemical J/2870/24
295.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Code : 23
Additional Details : Weight : 10.00000kg
Description
Buffer Solution pH9.2 (Borate), NIST Standard Solution ready to use for pH measurement
Specifications
Colorless | |
Liquid | |
163147 |
9.2 | |
10 L | |
Bag in box |