Learn More
Buffer Solution pH9.2 (Borate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical™
Brand: Fisher Chemical J/2870/15
64.04 GBP valid until 2024-03-30
Use promo code "21741" to get your promotional price.
Code : 23
Additional Details : CAS Number : 7732-18-5 Weight : 1.00000kg
Description
Buffer Solution pH9.2 (Borate), NIST Standard Solution ready to use for pH measurement
Specifications
Colorless | |
Liquid | |
163147 |
9.2 | |
1 L | |
HDPE plastic bottle |
Safety and Handling
- Danger
Hazard Category
- Reproductive toxicity Category 1B
Hazard Statement
- H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.