missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution pH8.00 (Borate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical™
Brand: Fisher Chemical J/2860/24
Code : 23
Additional Details : Weight : 10.00000kg
Description
Buffer Solution pH8.00 (Borate), NIST Standard Solution ready to use for pH measurement
Specifications
Colorless | |
Liquid | |
217672 |
8 | |
10 L | |
Bag in box |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Danger
Hazard Category
- Reproductive toxicity Category 1B
Hazard Statement
- H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.