Learn More
Buffer Solution pH7.00 (Phosphate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical™
Brand: Fisher Chemical J/2850/21
65.00 GBP valid until 2021-02-28
Use promo code "14802" to get your promotional price.
Code : 23
Additional Details : Weight : 5.00000kg
Packaging | HDPE plastic bottle |
---|---|
Quantity | 5L |
Description
Ready to use for pH measurement.Each solution is easily identified by its color, saving time and reducing errors.
Our buffer solutions are a high quality and reliable range of products that ensure accurate results for calibration/titration.
- Standard and color-coded buffer solutions
- Manufactured to a tolerance of ±0.02 pH units
- Accurately standardized and labelled with lot-specific pH at 20°C
- All buffers are traceable to NIST and BS 1647 standard
- Packed in HDPE containers to help protect the products integrity an ensure safe handling (refers to 1L, 2.5L, and 5L quantities)
- Standard buffers are available over the range pH2 to pH10
- Color-coded buffer solutions are available over the range pH4, pH7, and pH
Chemical Identifiers
217672 |
Specifications
217672 | |
HDPE plastic bottle | |
Colorless |
Liquid | |
7 | |
5L |
Special Offers
-
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.: 14802: 2021-02-28