Learn More
Buffer Solution pH6.00 (Phosphate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical™
Buffer Solution pH6.00 (Phosphate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical, Quantity: 1L, CAS: 7778-77-0, Colorless, Packaging: HDPE plastic bottle, pH: 6, MDL Number: 81857, Physical Form: Liquid
Brand: Fisher Chemical J/2840/15
43.79 GBP valid until 2021-04-30
Use promo code "14803" to get your promotional price.
Code : 23
Additional Details : CAS Number : 7732-18-5 Weight : 1.00000kg
Description
Buffer Solution pH6.00 (Phosphate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical, Quantity: 1L, CAS: 7778-77-0, Colorless, Packaging: HDPE plastic bottle, pH: 6, MDL Number: 81857, Physical Form: Liquid
Specifications
Sodium Hydroxide, Water, Dihydrogen Potassium Phosphate | |
Miscible | |
7778-77-0 | |
HDPE plastic bottle | |
Colorless |
Liquid | |
Buffer Solution | |
81857 | |
6 | |
1L |
Special Offers
-
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.: 14803: 2021-04-30