missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution pH10.00 (Borate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical™
167.76 GBP valid until 2024-12-31
Use promo code "21980" to get your promotional price.
Use promo code "21980" to get your promotional price.
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
Packaging:
HDPE plastic bottle
Plastic container
Description
Buffer Solution pH10.00 (Borate), NIST Standard Solution ready to use for pH measurement
Ready to use for pH measurement.
Each solution is easily identified by its color, saving time and reducing errors.
Our buffer solutions are a high quality and reliable range of products that ensure accurate results for calibration/titration.
Each solution is easily identified by its color, saving time and reducing errors.
Our buffer solutions are a high quality and reliable range of products that ensure accurate results for calibration/titration.
- Standard and color-coded buffer solutions
- Manufactured to a tolerance of ±0.02 pH units
- Accurately standardized and labelled with lot-specific pH at 20°C
- All buffers are traceable to NIST and BS 1647 standard
- Packed in HDPE containers to help protect the products integrity an ensure safe handling (refers to 1L, 2.5L, and 5L quantities)
- Standard buffers are available over the range pH2 to pH10
- Color-coded buffer solutions are available over the range pH4, pH7, and pH
Specifications
Color | Colorless |
pH | 10 |
Physical Form | Liquid |
Quantity | 5 L |
Packaging | Plastic container |
Safety and Handling
Signal Word
- Danger
- Reproductive toxicity Category 1B
- H360FD-May damage fertility. May damage the unborn child.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'