missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution, pH 9.21, METTLER TOLEDO™
Click to view available options
Quantity:
250 mL
6 x 250 mL
Specifications
Color | Blue |
pH | 9.21 |
Physical Form | Liquid |
Quantity | 6 x 250 mL |
Solution Type | pH Buffer |
Odor | None |
Packaging | 250mL |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'