missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution, pH 8.00, Reagecon™
Click to view available options
Quantity:
500 mL
1 L
pH:
8
Description
To ensure reliable, reportable pH measurements, this pH buffer standard is directly traceable to the IUPAC pH scale by an unbroken chain of traceability. Traceable analysis is necessary for consistency and universal acceptance of pH results, including acceptance by regulatory bodies.
- Traceable to SRM for NIST
- Certification to ISO 9001:2008
- Accredited to ISO 17025
Specifications
Melting Point | 0°C |
Boiling Point | 100°C |
Color | Colorless |
pH | 8 |
Physical Form | Liquid |
Quantity | 1 L |
Solution Type | pH Buffer |
Traceability to NIST | Yes |
Solubility Information | Fully miscible in water |
Safety and Handling
Product Identifier
- BUFFER SOLUTION pH 8.00 ± 0.01 a 25°C
- Danger
- Reproductive toxicity Category 1B
- H360-May damage fertility or the unborn child.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501-Dispose of contents/container to .
- MIXTURE LIST-Contains : boric acid, disodiumsalt
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'