missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution pH 4, Red, Reagecon™
Click to view available options
Quantity:
250 mL
500 mL
1 L
5 L
Packaging:
Bag-in-Box Container
HDPE Bottle GL45
HDPE Twin Neck Bottle
pH:
4
Description
- Colored
- Twin-neck containers with integrated calibrating chamber
- Tested at 20°C or 25°C
- Certified by ISO 17025 Accredited Test Method
- NIST traceable
- Presented in various pack sizes
Specifications
Color | Red |
pH | 4 |
Quantity | 500 mL |
Packaging | HDPE Bottle GL45 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'