Learn More
Buffer Solution, pH 3.5, Reagecon™
Manufactured in compliance with Ph. Eur. Chapter 4 (4.1.3). Reagecon™ Buffer Solution, pH 3.5 is tested at 20°C to an accuracy level of ±0.05. This calibration and control buffer is specially formulated to ensure its stability, and is ISO 17025 accredited and NIST traceable.
Brand: Reagecon 4000600
Code : A0
Additional Details : Weight : 0.93400kg Transport : UN number : 1789 Chem class : 8 Pack group : II
Description
To ensure reliable, reportable pH measurements, this pH buffer standard is directly traceable to the IUPAC pH scale by an unbroken chain of traceability. Traceable analysis is necessary for consistency and universal acceptance of pH results, including acceptance by regulatory bodies.
- Manufactured and tested in compliance with the Ph. Eur. Chapter 4 (4.1.3)
- Supplied with a detailed certificate of analysis, which outlines traceability to NIST
- Certification to ISO 9001:2008 and accredited to ISO 17025
- Temperature dependence data is printed on label, as are lot numbers and expiry dates
- Extended shelf life with enhanced stability—usable to expiry date, even after opening
Specifications
0°C | |
Colorless | |
3.5 ±0.05 | |
1 L | |
Yes |
100°C | |
Yes | |
Liquid | |
pH Buffer | |
Fully miscible in water |
Safety and Handling
- BUFFER SOLUTION pH 3.5
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : hydrochloric acid 37%