missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution, pH 2.00 (+/-0.01 @ 25oC), No Color, Specpure, NIST Traceable, Thermo Scientific Chemicals

For Calibration of pH Meters

Brand:  Thermo Scientific Alfa Aesar 040447.K2

Code : NEW

Additional Details : CAS Number : 7647-01-0 Weight : 0.01000kg

 View more versions of this product

Product Code. 11358408

  • £30.10 / 1 L
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Buffer Solution pH 2 used as a means of keeping pH at a nearly constant value in a different chemical applications. Glycine Buffer Solution pH 2 used for eluting antibodies in various antibody purification procedures i.e. protein A-agarose, protein G-agarose, affinity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Buffer solution
Colorless
Liquid
MFCD00146252
23 hPa (17mm Hg) at 20°C
Potassium chloride: 0.7%; Hydrochloric acid: 0.7%; water: 98.6%
7447-40-7,7647-01-0,7732-18-5
1.005g/cm3 at 20°C
1 L
Yes
Soluble in water.
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only