missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Buffer solution, pH 13.00 (+/-0.01 at 25°C), No Color, Specpure™, NIST Traceable

Brand:  Alfa Aesar™ 046307.K7

Code : NEW

Additional Details : Weight : 4.00000kg

Quantity 4L

 View more versions of this product

Product Code. 15474035

  • £37.40 / 4 L


Estimated Shipment date
from Supplier 25-04-2018
Add to basket

Specifications

Chemical Identifiers

Buffer solution, pH 13.00 (±0.01 at 25°C)
NIST Traceable
Specpure™
MFCD03458582

Specifications

Buffer solution, pH 13.00 (±0.01 at 25°C)
Specpure™
MFCD03458582
4L
NIST Traceable
Liquid
Colorless
Videos
SDS