missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Color Coded pH Buffer Solution Tested at 25°C, Reagecon™
Click to view available options
Quantity:
500 mL
1 L
5 L
Packaging:
Bag-in-Box Container
HDPE Bottle
HDPE Bottle GL45
Color:
Blue
Yellow
Description
- Colored pH Buffer Solutions
- Tested at 25°C
- Certified by Reagecon’s ISO/IEC 17025 AccreditedTest Method
- NIST traceable
- Presented in various pack sizes
Specifications
Color | Blue |
pH | 10 |
Physical Form | Liquid |
Quantity | 500 mL |
Packaging | HDPE Bottle |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'