Learn More
Buffer Solution, pH 10, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 258600025
Code : 29
Additional Details : CAS Number : 00-00-0 Weight : 2.55000kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
pH bufferSpecifications
Buffer solution pH 10 | |
1332-57-6,144-55-8,7732-18-5 | |
Liquid | |
Plastic bottle |
Colorless | |
9.98 to 10.02 (20°C) | |
2.5 L | |
Solubility in water: soluble. |
Safety and Handling
- Buffer solution pH 10, Carbonate buffer
RUO – Research Use Only