Learn More
Buffer solution, pH 1.68 (+/-0.026 @ 25°C), No Color, Specpure, NIST Traceable, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 042415.AP
14.40 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : CAS Number : 7732-18-5 Weight : 0.01000kg
Description
Buffer Solution pH 1.68 used as a means of keeping pH at a nearly constant value in a different chemical applications. Glycine buffer solution pH 2 used for eluting antibodies in various antibody purification procedures i.e. protein A-agarose, protein G-agarose, affinity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Buffer solution | |
Colorless | |
Liquid | |
Odorless | |
23 hPa (17mm Hg) at 20°C | |
Potassium chloride: 0.7%; Hydrochloric acid: 0.7%; water: 98.6% |
7447-40-7,7647-01-0,7732-18-5 | |
1.005g/cm3 at 20°C | |
500 mL | |
MFCD00147253 | |
Miscible with water. |
RUO – Research Use Only