Learn More
Buffer solution, pH 1.00 (+/-0.026 @ 25oC), No Color, Specpure, NIST Traceable, Thermo Scientific Chemicals
For Calibration of pH Meters
Brand: Thermo Scientific Alfa Aesar 040446.K2
Code : NEW
Additional Details : CAS Number : 7447-40-7 Weight : 0.01000kg
Description
Calibration of pH meters
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Buffer solution | |
Colorless | |
Liquid | |
MFCD00146252 | |
Potassium chloride: 0.7%; Hydrochloric acid: 0.7%; water: 98.6% |
7447-40-7,7647-01-0,7732-18-5 | |
1.005 | |
1 L | |
23 hPa (17mm Hg) at 20°C |
RUO – Research Use Only