missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution for HPCE, 20 mM sodium phosphate, pH 7.5 (25 °C), Honeywell Fluka™

Brand:  Honeywell Fluka 82592-500ML

Code : NEW

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 15637210

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Specifications

1: 0 to 0.025; 2: 0 to 0.07; 3: 0 to 0.6
UV absorption/200 to 800nm in accordance
Liquid
20mM sodium phosphate
Plastic bottle
500mL
Buffer solution HPCE pH 7.5
Colorless
2 to 2.7mS/cm at 20°C
NONH for all modes of transport
7.5 (25°C)
In accordance for HPCE
Videos