missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution, Di-Sodium Tetraborate/Sodium Hydroxide, Honeywell™
Click to view available options
Quantity:
1 L
Specifications
CAS | 1336-21-6 |
Density | 1.0g/mL at 20°C |
Quantity | 1 L |
Molecular Formula | H5NO |
MDL Number | MFCD00066650 |
UN Number | NONH for all modes of transport |
InChI Key | VHUUQVKOLVNVRT-UHFFFAOYSA-N |
SMILES | N.O |
IUPAC Name | amine hydrate |
Molecular Weight (g/mol) | 35.05 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'