missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Fluka™ Buffer solution pH 4.0 (20°C), Red colored, citric acid / sodium hydroxide / sodium chloride solution, with fungicide, Honeywell Fluka™

Red colored, citric acid / sodium hydroxide / sodium chloride solution, with fungicide

£19.97 - £183.75

Chemical Identifiers

CAS 1336-21-6
Molecular Formula H5NO
Molecular Weight (g/mol) 35.046
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
PubChem CID 14923
ChEBI CHEBI:18219
IUPAC Name azanium;hydroxide
SMILES [NH4+].[OH-]
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15654060 View Documents Honeywell Fluka™
33665-500ML
500mL £19.97
500mL View alternative products
Save Up to 
»
In Stock 2
Add to basket
 
15684060 View Documents Honeywell Fluka™
33665-6X500ML
6 x 500mL £106.38
Pack of 6 View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 25-02-2020
Add to basket
 
15674060 View Documents Honeywell Fluka™
33665-6X1L
6 x 1L £175.44
Pack of 6 View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 25-02-2020
Add to basket
 
15644060 View Documents Honeywell Fluka™
33665-1L
1L £33.81
1 L View alternative products
Save Up to 
»
In Stock 6
EU Stock 3
Add to basket
 
15664060 View Documents Honeywell Fluka
33665-5L-VP
5L £76.42
5 L View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Add to basket
 
15634060 View Documents Honeywell Fluka
33665-10L-VP
10L £183.75
10 L View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Add to basket
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

1336-21-6
35.046
14923
azanium;hydroxide
H5NO
VHUUQVKOLVNVRT-UHFFFAOYSA-N
CHEBI:18219
[NH4+].[OH-]

Specifications

Buffer solution
H5NO
VHUUQVKOLVNVRT-UHFFFAOYSA-N
azanium;hydroxide
14923
4.0 (20°C)
1336-21-6
NONH for all modes of transport
[NH4+].[OH-]
35.046
CHEBI:18219
500mL