missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution pH 4.0 (20°C), Red colored, citric acid / sodium hydroxide / sodium chloride solution, with fungicide, Honeywell Fluka™

Catalog Number 15654060
Click to view available options
Quantity:
500 mL
1 L
5 L
6 x 500 mL
10 L
6 x 1 L
This item is not returnable. View return policy
1336-21-6
H5NO
35.05
MFCD00066650
VHUUQVKOLVNVRT-UHFFFAOYSA-N
14923
CHEBI:18219
N.O
This item is not returnable. View return policy
1336-21-6
H5NO
35.05
MFCD00066650
VHUUQVKOLVNVRT-UHFFFAOYSA-N
14923
CHEBI:18219
N.O

Red colored, citric acid / sodium hydroxide / sodium chloride solution, with fungicide

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
pH 4.0
Quantity 500 mL
UN Number NONH for all modes of transport
IUPAC Name azanium;hydroxide
Chemical Name or Material Buffer solution
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'