missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Colour Coded Solution pH7.00 (Phosphate) Yellow, NIST Standard Solution ready to use for pH measurement, Fisher Chemical™
39.75 GBP valid until 2024-12-31
Use promo code "21980" to get your promotional price.
39.75 GBP valid until 2024-03-30
Use promo code "21741" to get your promotional price.
50.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "21980" to get your promotional price.
39.75 GBP valid until 2024-03-30
Use promo code "21741" to get your promotional price.
50.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
10 L
Packaging:
Bag in box
HDPE plastic bottle
Plastic container
Description
Buffer Colour Coded Solution pH7.00 (Phosphate) Yellow, NIST Standard Solution ready to use for pH measurement, Fisher Chemical, Quantity: 1L, Packaging: HDPE plastic bottle, pH: 7, MDL Number: 81857, Physical Form: Liquid
Ready to use for pH measurement.
Each solution is easily identified by its color, saving time and reducing errors.
Our buffer solutions are a high quality and reliable range of products that ensure accurate results for calibration/titration.
Each solution is easily identified by its color, saving time and reducing errors.
Our buffer solutions are a high quality and reliable range of products that ensure accurate results for calibration/titration.
- Standard and color-coded buffer solutions
- Manufactured to a tolerance of ±0.02 pH units
- Accurately standardized and labelled with lot-specific pH at 20°C
- All buffers are traceable to NIST and BS 1647 standard
- Packed in HDPE containers to help protect the products integrity an ensure safe handling (refers to 1L, 2.5L, and 5L quantities)
- Standard buffers are available over the range pH2 to pH10
- Color-coded buffer solutions are available over the range pH4, pH7, and pH
Specifications
Color | Yellow |
pH | 7 |
Physical Form | Liquid |
Quantity | 1 L |
Solution Type | Buffer |
Formulation | Dihydrogen Potassium Phosphate, Sodium Chloride, Water, Sodium Phosphate Dibasic |
MDL Number | MFCD00011401 MFCD00147253 |
Packaging | HDPE plastic bottle |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'