missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Capsules pH 7, Reagecon™
Click to view available options
Quantity:
50 Capsules
Description
- Colour coded for ease of identification
- Easy to use
- Dissolve quickly
- Accuracy ±0.02 pH units
- Preservative free
- Economical
- Easy to store and transport
- Extended shelf life
Specifications
Color | Green |
pH | 7 |
Quantity | 50 Capsules |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'