missing translation for 'onlineSavingsMsg'
Learn More

Budesonide, 97%, Thermo Scientific Chemicals

Catalog Number 15539533
95.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
51333-22-3
C25H34O6
430.54
MFCD00083259,MFCD00138154
VOVIALXJUBGFJZ-VXKMTNQYSA-N
73759663
(1S,2S,4R,6R,8S,9S,11S,12S,13R)-11-hydroxy-8-(2-hydroxyacetyl)-9,13-dimethyl-6-propyl-5,7-dioxapentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icosa-14,17-dien-16-one
CCC[C@@H]1O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@H]4[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO
51333-22-3
C25H34O6
430.54
MFCD00083259,MFCD00138154
VOVIALXJUBGFJZ-VXKMTNQYSA-N
73759663
(1S,2S,4R,6R,8S,9S,11S,12S,13R)-11-hydroxy-8-(2-hydroxyacetyl)-9,13-dimethyl-6-propyl-5,7-dioxapentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icosa-14,17-dien-16-one
CCC[C@@H]1O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@H]4[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol and chloroform, slightly soluble in acetone, ethanol, acetonitrile, very slightly soluble in ether, dichloromethane
Formula Weight 430.54
Percent Purity 97%
Chemical Name or Material Budesonide
compliance-icons compliance-icons
Product Identifier
  • Budesonide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H360-May damage fertility or the unborn child.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'