Learn More
Eppendorf™ Buckets for Rotor S-4x1000
Centrifuge Buckets for Rotor S-4x1000 are available in a variety of sizes. Offered in packs of two and four.
Brand:  Eppendorf™ 5895104002
Additional Details : Weight : 4.23000kg
 View more versions of this product
Product Code. 15718369
  • £1875.00 / Pack of 4
Estimated Shipment: 17-05-2024
to see stock.
Description
Description
A variety of centrifuge buckets to fit your needs:
  • Round bucket S-4x1000 4-piece set
  • Round bucket S-4x1000 2-piece set
  • Plate/tube bucket S-4x1000 4-piece set
  • Plate/tube bucket S-4x1000 2-piece set
  • High-capacity bucket S-4x1000 4piece set
  • High-capacity bucket S-4x1000 2-pieces
Specifications
Specifications
Rotor S-4x1000
Plate/Tube Bucket
Set of 4 pcs.