missing translation for 'onlineSavingsMsg'
Learn More

Buchi Evaporation tubes graduated working volume 25 to 250mL residual volume 1mL

Brand:  Buchi 038575

Evaporation tubes graduated working volume 25 to 250mL residual volume 1mL

Code : 62

Additional Details : Weight : 1.73000kg

Product Code. 12826713

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.