missing translation for 'onlineSavingsMsg'
Learn More

BUCHI™ Syncore™ Glass Tube Set

Brand:  BUCHI™ 038557

Code : 62

Additional Details : Weight : 1.94000kg

 View more versions of this product

Product Code. 12896703

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Specifications

Graduated glass Tube; Residual volume 0.3mL
0 to 600rpm
250mL
Videos