missing translation for 'onlineSavingsMsg'
Learn More

BUCHI™ Receiving Flasks for Rotary Evaporators

Use these receiving flasks with any rotary evaporator. Buchi™ Spherical Receiving Flasks offer high chemical resistance against acids, alkalis and organic substances. Ideal for collecting condensed solvents.

Brand:  BUCHI™ 020728

Code : A0

Additional Details : Weight : 0.37600kg

 View more versions of this product

Product Code. 10489492

  • £160.00 / Each


In Stock 3
Add to basket

Description

Description

  • Spherical Receiving Flasks for rotary evaporators with ball joint (35/20)
  • Exclusive use of DURAN borosilicate glass 3.3
  • High chemical resistance against acids, alkalis and organic substances
  • Resistant to thermal shocks and high temperatures combined with low thermal expansion
  • Constant monitoring of glass wall thickness uniformity
  • High degree of attention paid to sphericity of rotating glass parts
  • Glass parts are tension-relieved at 560°C
  • Available in volumes from 50mL to 3000mL
  • Available uncoated or with P+G coating

Specifications

Specifications

1000mL
Receiving Flask
Ball joint KS-35
Rotary evaporator
Plastic and Glass
Round
Videos
Documentation