missing translation for 'onlineSavingsMsg'
Learn More

BUCHI™ Capillaries for Melting Point Apparatus

For use with Buchi Melting Point apparatus.

Brand:  BUCHI™ 001759

UNSPSC : 41112203

Code : 61

Additional Details : Weight : 0.36000kg

 View more versions of this product

Product Code. 12847685

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

For use with Buchi Melting Point apparatus.
Specifications

Specifications

Melting Point Capillary
Laboratory general purpose tubing
Glass
1000 tubes