missing translation for 'onlineSavingsMsg'
Learn More

BUCHI™ Large Capacity Evaporating Flasks

Standard glass flasks for use with various Rotavapor™ Evaporator models

Brand:  BUCHI™ 011899

Code : 62

Additional Details : Weight : 3.45000kg

 View more versions of this product

Product Code. 12782405

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Specifications

20L
R-151 rotary evaporator
Standard Glass
20000mL
Evaporating Flask
Videos