missing translation for 'onlineSavingsMsg'
Learn More

BUCHI™ Large Capacity Evaporating Flasks

Standard glass flasks for use with various Rotavapor™ Evaporator models

Brand:  BUCHI™ 027470

Code : X3

Additional Details : Weight : 1.93000kg

 View more versions of this product

Product Code. 10267813

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

  • Inert industrial glass (borosilicate 3.3, ISO/DIN 3585)
  • Tension relieved at 540°C

Specifications

Specifications

20L
R-153 and R-220 rotary evaporator
Standard Glass
6000mL
Evaporating Flask
Videos