Learn More
BUCHI™ Glass Precolumns
Brand:  BUCHI™ 044026
UNSPSC : 41115700
Code : 62
Additional Details : Weight : 0.19000kg
 View more versions of this product
Product Code. 12795985
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Specifications
Specifications
Glass column with I.D. 15, 26, 36 and 49mm
Glass
Precolumn
HPLC
Documentation