missing translation for 'onlineSavingsMsg'
Learn More

Buchi Plunger column 36/460 biocompatable with integrated cooling jacket I.D. 36mm x length 460mm

Brand:  Buchi 044063

Plunger column 36/460 biocompatable with integrated cooling jacket I.D. 36mm x length 460mm

Code : 62

Additional Details : Weight : 3.02000kg

Product Code. 12729495

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.