missing translation for 'onlineSavingsMsg'
Learn More

Buchi Glass column 15/460 cpl Plastic+Glas coated for preparative chromatography ID 15mm x length 460mm

Brand:  Buchi 044032

Glass column 15/460 cpl Plastic+Glas coated for preparative chromatography ID 15mm x length 460mm

Code : 62

Additional Details : Weight : 1.33000kg

Product Code. 12735995

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.