missing translation for 'onlineSavingsMsg'
Learn More

BUCHI™ Glass Columns C-690

Brand:  BUCHI™ 044044

Code : 62

Additional Details : Weight : 1.53000kg

 View more versions of this product

Product Code. 12729485

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Specifications

Preparative separations together with the C-600 Sepacore™ line of chromatography separators
Borosilicate Glass
230mm
290psi
Glass column
49mm
Videos
Documentation View accessory products