missing translation for 'onlineSavingsMsg'
Learn More

BUCHI™ Glass Columns C-690

Brand:  BUCHI™ 044040

Code : 62

Additional Details : Weight : 1.61000kg

 View more versions of this product

Product Code. 12799475

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

Preparative separations together with the C-600 Sepacore™ line of chromatography separators
Borosilicate Glass
230mm
435psi
Glass column
36mm
Videos
Documentation