missing translation for 'onlineSavingsMsg'
Learn More

Buchi Glass column 100/230 cpl Plastic+Glas coated for preparative chromatography ID 100mm x length 230mm

Brand:  Buchi 044052

Glass column 100/230 cpl Plastic+Glas coated for preparative chromatography ID 100mm x length 230mm

Code : 62

Additional Details : Weight : 4.85000kg

Product Code. 12789485

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.