missing translation for 'onlineSavingsMsg'
Learn More

Buchi Glass column 70/230 cpl Plastic+Glas coated for preparative chromatography ID 70mm x length 230mm

Brand:  Buchi 044048

Glass column 70/230 cpl Plastic+Glas coated for preparative chromatography ID 70mm x length 230mm

Code : 62

Additional Details : Weight : 2.98000kg

Product Code. 12759485

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.