missing translation for 'onlineSavingsMsg'
Learn More

Buchi Glass column 15/920 cpl Plastic+Glas coated for preparative chromatography ID 15mm x length 920mm

Brand:  Buchi 044033

Glass column 15/920 cpl Plastic+Glas coated for preparative chromatography ID 15mm x length 920mm

Code : 62

Additional Details : Weight : 1.60000kg

Product Code. 12745995

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.