missing translation for 'onlineSavingsMsg'
Learn More

Buchi Tube glass extension 460mm x 70mm

Brand:  Buchi 017979

Tube glass extension 460mm x 70mm

Code : 62

Additional Details : Weight : 1.34000kg

Product Code. 12719535

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.