missing translation for 'onlineSavingsMsg'
Learn More

Buchi Tube glass extension 460mm x 26mm

Brand:  Buchi 017982

Tube glass extension 460mm x 26mm

Code : 62

Additional Details : Weight : 0.30000kg

Product Code. 12789525

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.