missing translation for 'onlineSavingsMsg'
Learn More

BUCHI™ Evaporating Flask and Accessories

Flasks made of borosilicate 3.3 glass which is resistant to chemicals and temperature changes of 180°C

Brand:  BUCHI™ 000779

Code : 61

Additional Details : Weight : 0.06000kg

 View more versions of this product

Product Code. 12752385

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

100mL
Evaporating Flask
180°C
Evaporation system
Borosilicate Glass
Videos