missing translation for 'onlineSavingsMsg'
Learn More

BUCHI™ Beaker-Style Drying Flasks for Rotary Evaporators

Brand:  BUCHI™ 034269

Code : A0

Additional Details : Weight : 0.72000kg

 View more versions of this product

Product Code. 10657892

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

500mL
Drying Flask
Plastic and Glass
60°C
Rotary evaporator
Standard Glass
Pear
29.2/32
Videos
Documentation