BUCHI™ Beaker Flasks with Flange
Overview
Brand: BUCHI™
Manufacturer Part Number: 034142
Beaker Flask With Flange
UNSPSC: 41121804
Code: 61
Additional Details:
Additional Details: Weight: 0.74000kg
Product Code. 12722345
Quantity Price
1 £ 516.77 / Each
Estimated Shipment
02-03-2017
Add to basket
Description and Specification
Specification
Product Type Beaker Flask
Capacity (Metric) 500mL
For Use With (Equipment) R-200 and R-205 rotary evaporator
Includes Flange