Learn More
BUCHI Boiling Point Tubes and Capillaries
Boiling point tubes and capillaries for automated or manual determination according to the Siwoloboff method.
Brand: BUCHI 019007
Code : A0
Additional Details : Weight : 0.11000kg
Description
- High precision for reliable resutls
- Meets Siwoloboff and Eu Pharmacopeia standards
- Designed for BUCHI melting and boiling point instruments
- Compatible with other instruments
Specifications
Boiling Point Tubes | |
For reproducible boiling point determinations | |
Borosilicate Glass 3.3 | |
Autoclavable |
2.75 mm | |
Boiling point apparatus | |
100/Pk. |