Learn More
BUCHI Boiling Point Tubes and Capillaries
Boiling point tubes and capillaries for automated or manual determination according to the Siwoloboff method.
Brand:  BUCHI 051890
Code : 61
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 12201507
  • £810.00 / Pack of 100
Estimated Shipment: 29-05-2024
to see stock.
Add to basket
Description
Description
  • High precision for reliable resutls
  • Meets Siwoloboff and Eu Pharmacopeia standards
  • Designed for BUCHI melting and boiling point instruments
  • Compatible with other instruments
Specifications
Specifications
Boiling Point Capillaries
Laboratory general purpose tubing
Borosilicate Glass 3.3
Autoclavable
0.8 mm
Boiling point apparatus
100/Pk.